Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14622

Sigma-Aldrich

4-Methoxybenzoic acid sodium salt

≥9% (HPLC)

Synonym(s):

4-Methoxybenzoic acid sodium salt, p-Anisic acid sodium salt, Sodium p-anisate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NaO3
CAS Number:
Molecular Weight:
174.13
UNSPSC Code:
12352107
EC Index Number:
208-634-1

form

powder

concentration

≥9% (HPLC)

storage temp.

2-30°C

InChI

1S/C8H8O3.Na/c1-11-7-4-2-6(3-5-7)8(9)10;/h2-5H,1H3,(H,9,10);/q;+1/p-1

InChI key

AETSDHMVQHOYPB-UHFFFAOYSA-M

General description

for synthesis

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 98
Purity (HPLC, area%): ≥ 98
Identity (IR): passes test
Water (K. F.): ≤ 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service