Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14499

Sigma-Aldrich

Dimethyl adipate/Dimethyl glutarate/Dimethyl succinate

(mixture of the esters 16:63:21) for synthesis

Synonym(s):

Imsol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

form

liquid

expl. lim.

0.9-8.0 % (v/v)

mp

-20 °C

transition temp

flash point 102 °C

solubility

53 g/L

density

1.09 g/cm3 at 20 °C

Storage temp.

2-30°C

Analysis Note

Assay cpmplete test mixture (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.086 - 1.091
Identity (IR): passes test

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

215.6 °F

flash_point_c

102 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service