Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14018

Sigma-Aldrich

Tetrakis(diethylamino)titanium

for synthesis

Synonym(s):

Tetrakis(diethylamino)titanium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H40N4Ti
CAS Number:
Molecular Weight:
336.38
MDL number:
UNSPSC Code:
12352103

Quality Level

form

liquid

bp

85-90 °C/0.1 hPa

transition temp

flash point 10 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/4C4H10N.Ti/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

VJDVOZLYDLHLSM-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 0.929 - 0.931
Identity (IR): passes test

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class

4.3 - Hazardous materials, which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

-19.8 °F - closed cup

flash_point_c

-28.8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service