Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03505

Sigma-Aldrich

4-Heptanone

for synthesis

Synonym(s):

4-Heptanone, Dipropyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2CO
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-608-9
Product 8.03505 is not currently sold in your country. Contact Technical Service

vapor pressure

7 hPa ( 20 °C)

Quality Level

assay

≥98.0% (GC)

form

liquid

autoignition temp.

430 °C

potency

3021 mg/kg LD50, oral (Rat)
4585 mg/kg LD50, skin (Rabbit)

expl. lim.

1 % (v/v)

mp

-33 °C

transition temp

flash point 49 °C

solubility

3.2 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H14O/c1-3-5-7(8)6-4-2/h3-6H2,1-2H3

InChI key

HCFAJYNVAYBARA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.813 - 0.815
Identity (IR): passes test

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service