Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01522

Sigma-Aldrich

Dibenzyl ether

for synthesis

Synonym(s):

Dibenzyl ether, Benzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-118-2

vapor pressure

<1 Pa ( 25 °C)

Quality Level

assay

≥98.0% (GC)

form

liquid

autoignition temp.

470 °C

potency

2500 mg/kg LD50, oral (Rat)

bp

298 °C/1013 hPa

mp

4 °C

transition temp

flash point 137 °C

solubility

insoluble 42 mg/L

density

1.04 g/cm3 at 20 °C

InChI

1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MHDVGSVTJDSBDK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.043 - 1.044
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

278.6 °F - Pensky-Martens closed cup

flash_point_c

137 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service