Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00678

Sigma-Aldrich

Succinyl dichloride

for synthesis

Synonym(s):

Succinyl dichloride, Succinic acid dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$156.00

$156.00


Check Cart for Availability


Select a Size

Change View
50 ML
$156.00

About This Item

Empirical Formula (Hill Notation):
C4H4Cl2O2
CAS Number:
Molecular Weight:
154.98
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-838-0

$156.00


Check Cart for Availability

Quality Level

mp

20 °C

transition temp

flash point 76 °C

density

1.38 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)CCC(=O)Cl

InChI

1S/C4H4Cl2O2/c5-3(7)1-2-4(6)8/h1-2H2

InChI key

IRXBNHGNHKNOJI-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 95.0 %
Density (d 20 °C/ 4 °C): 1.375 - 1.385
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible, corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service