Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

7410-OP

Millipore

D-(-)-Ribose

OmniPur® Grade, >=99.0%

Synonym(s):

OmniPur® D-(-)-Ribose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

wet ice

InChI

1S/C5H10O5/c6-1-2-3(7)4(8)5(9)10-2/h2-9H,1H2

InChI key

HMFHBZSHGGEWLO-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
Arsenic: (As) ≤0.0002%
Ash: ≤0.2%
Assay: ≥98.0%
Heavy metals (as Pb): ≤0.001%
Moisture (H2O): ≤3.0%
Specific rotation [a]20/D: -21--19°





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service