Key Documents
71184-M
Sigma-Aldrich
His•Tag® Antibody Plate
Immobilized antibody in 96-well format
About This Item
UNSPSC Code:
12352203
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
storage temp.
2-8°C
General description
The His•Tag Antibody Plate is a 96-well, ELISA-compatible plate containingimmobilized His•Tag Monoclonal Antibody.The antibody is covalently immobilized to thesurface using a method that retains maximalbinding activity. The antibody specificallyrecognizes five consecutive histidines, and sowill bind with high affinity (Kd = 5 × 10-8 to1 × 10-9 M) to virtually any His•Tag fusionprotein in which the tag is exposed. Thisplate has outstanding binding characteristics,with a capacity of >100 ng His•Tag fusionprotein per well and low non-specific binding.Well-to-well variability is less than 5% andthe plates are stable when stored dry at 4°C.The His•Tag Antibody Plate can be used ina variety of binding assays where reliable,specific immobilization of His•Tag fusionproteins is required.
Warning
Toxicity: Standard Handling (A)
Legal Information
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany