Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

684500

Sigma-Aldrich

Zaprinast - CAS 37762-06-4 - Calbiochem

A cell-permeable, selective inhibitor of cGMP-specific phosphodiesterase (PDE V; IC₅₀ = 450 nM).

Synonym(s):

Zaprinast - CAS 37762-06-4 - Calbiochem, 1,4-Dihydro-5-(2-propoxyphenyl)-7H-1,2,3-triazolo[4,5-d]pyrimidine-7-one, M&B22948

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N5O2
CAS Number:
Molecular Weight:
271.27
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

potency

450 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

tan to pink

solubility

DMSO: 25 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C13H13N5O2/c1-2-7-20-9-6-4-3-5-8(9)11-14-12-10(13(19)15-11)16-18-17-12/h3-6H,2,7H2,1H3,(H2,14,15,16,17,18,19)

InChI key

REZGGXNDEMKIQB-UHFFFAOYSA-N

General description

A cell-permeable, selective inhibitor of cGMP-specific phosphodiesterase (PDE V; IC50 = 450 nM). Inhibits PDE IX at higher concentration (IC50 = 35 µM). Also known to increase cGMP levels in spontaneously hypertensive rat (SHR) plasma and reverse nitroglycerin tolerance in vitro. Enhances the vasodilatory effects of nitric oxide.
Selective inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50 = 450 nM). Also known to increase cGMP levels in SHR plasma and reverse nitroglycerin tolerance in vitro. Enhances the vasodilatory effects of nitric oxide.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cGMP-specific phosphodiesterase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.







From Catalog:

Desc. Field- added "cell-permeable"

Other Notes

Fisher, D.A., et al. 1998. J. Biol. Chem. 273, 15559.
Tsusu, K.G., et al. 1995. Am. J. Respir. Crit. Care Med.152, 1605.
Dundore, R.L., et al. 1993. Eur. J. Pharmacol.249, 293.
Pagani, E.D., et al. 1993. Eur. J. Pharmacol.243, 141.
Burns, F., et al. 1992. Biochem. J.283, 487.
Lugnier, C., et al. 1992. Am. J. Physiol.262, H654.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice L Herneisen et al.
eLife, 13 (2024-08-13)
Apicomplexan parasites balance proliferation, persistence, and spread in their metazoan hosts. AGC kinases, such as PKG, PKA, and the PDK1 ortholog SPARK, integrate environmental signals to toggle parasites between replicative and motile life stages. Recent studies have cataloged pathways downstream
H Ji
Methods in molecular medicine, 51, 363-377 (2001-01-01)
Angiotensin type-1 receptors (AT(1) receptors) mediate various physiological actions of angiotensin (Ang II) via multiple-signal transduction pathways (1). In addition to the phospholipase C pathway and dihydropyridine-sensitive voltage-dependent calcium channels, AT(1) receptors can couple to inhibition of adenylate cyclase via
Alex W Chan et al.
eLife, 12 (2023-11-07)
Apicomplexan parasites use Ca2+-regulated exocytosis to secrete essential virulence factors from specialized organelles called micronemes. Ca2+-dependent protein kinases (CDPKs) are required for microneme exocytosis; however, the molecular events that regulate trafficking and fusion of micronemes with the plasma membrane remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service