681666
InhibitorSelect Wnt Signaling Pathway Inhibitor Panel
The InhibitorSelect Wnt Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.
About This Item
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
protect from light
solubility
DMSO: soluble (supplied)
shipped in
ambient
storage temp.
−20°C
General description
Packaging
Warning
Reconstitution
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
188.6 °F - (refers to pure substance)
flash_point_c
87 °C - (refers to pure substance)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service