Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681637

Sigma-Aldrich

Wee1/Chk1 Inhibitor - CAS 1177150-89-8 - Calbiochem

The Wee1/Chk1 Inhibitor, also referenced under CAS 1177150-89-8, controls the biological activity of Wee1/Chk1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Wee1/Chk1 Inhibitor - CAS 1177150-89-8 - Calbiochem, 4-(2-Phenyl)-9-hydroxypyrrolo[3,4-c]carbazole-1,3-(2H,6H)-dione, Chk1 Inhibitor IV, Wee1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N2O3 · xH2O
CAS Number:
Molecular Weight:
328.32 (anhydrous basis)
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 200 mg/mL
ethanol: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A pyrrolocarbazole compound that acts as a potent and ATP-competitive inhibitor of checkpoint kinases Wee1 and Chk1 (IC50 = 97 nM and 47 nM, respectively). Inhibits PKC, Cdk4, or other CDKs only at much higher concentrations (IC50 = 3.4 µM, 3.75 µM, and >5 µM, respectively) and exhibits little effect against FGFR, PDGFR, or c-Src (IC50 >50 µM).
A pyrrolocarbazole compound that acts as a potent and ATP-competitive inhibitor of checkpoint kinases Wee1 and Chk1 (IC50 = 97 nM and 47 nM, respectively). Inhibits PKC, Cdk4, or other CDKs only at much higher concentrations (IC50 = 3.4 µM, 3.75 µM, and >5 µM, respectively) and exhibits little effect against FGFR, PDGFR, or c-Src (IC50 >50µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Wee1 and Chk1
Product competes with ATP.
Reversible: no
Target IC50: 97 nM and 47 nM against Wee1 and Chk1, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Palmer, B.D., et al. 2006. J. Med. Chem.49, 4896.
Squire, C.J., et al. 2005. Structure13, 541.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lionel Condé et al.
BMC molecular and cell biology, 22(1), 16-16 (2021-03-06)
Staufen2 (STAU2) is an RNA binding protein involved in the posttranscriptional regulation of gene expression. In neurons, STAU2 is required to maintain the balance between differentiation and proliferation of neural stem cells through asymmetric cell division. However, the importance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service