Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681105-M

Sigma-Aldrich

Vitronectin, Human Plasma

Synonym(s):

Serum Spreading Factor, S-Protein

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥95% (SDS-PAGE)

form

lyophilized

solubility

aqueous buffer: soluble
distilled water: soluble

General description

Intact protein and SDS-PAGE bands react with monoclonal antibodies to vitronectin, bind to heparin, and promote cell adhesion.
M.W. 65,000 and 75,000.
Native vitronectin purified from human plasma. Extracellular matrix protein that promotes cell adhesion. Exists as a mixture of 75 kDa and 65 kDa polypeptides. Biochemically and immunologically distinct from fibronectin. Binds to cells through the RGD sequence in its cell-binding domain.
Native vitronectin purified from human plasma. Extracellular matrix protein that promotes cell adhesion. Exists as a mixture of ~75 kDa and ~65 kDa polypeptides. Biochemically and immunologically distinct from fibronectin. Intact protein and SDS-PAGE bands react with monoclonal antibodies to vitronectin, bind to heparin, and promote cell adhesion.

Preparation Note

Equilibrate vial to room temperature prior to reconstitution. Swirl gently to dissolve.
Prepared from plasma of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Hayman et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(13), 4003-4007 (1983-07-01)
Monoclonal antibodies were prepared against a cell attachment-promoting protein, serum spreading factor, which had been partially purified from human serum by chromatography on glass bead columns. The antibodies selected were those that reacted with polypeptides that had cell attachment-promoting activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service