Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

65067-M

Sigma-Aldrich

Hematoxylin Gill III

Harleco (For Microscopy (contains ethylene glycol, Aluminum sulfate hexadecahydrate, acetic acid, Sodium iodate))

Synonym(s):

H&E stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

form

liquid

manufacturer/tradename

Harleco®

mp

16.67 °C

density

1.16 g/cm3

λmax

545-560 nm

application(s)

diagnostic assay manufacturing
hematology

storage temp.

15-25°C

General description

HARLECO® Hematoxylin
65067-75 1L Poly Bottle
HARLECO® Triple strength nuclear stain used for routine tissue specimens glycol and methylmethacrylate sections. For regressive staining

Application

HARLECO® Triple strength nuclear stain used for routine tissue specimens glycol and methylmethacrylate sections. For regressive staining
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Same as Gill I but triple the strength. Can be used for routine tissue specimens glycol and methylmethacrylate sections. For regressive staining.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service