Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65065-M

Sigma-Aldrich

Hematoxylin Gill I

Harleco® (For Microscopy (contains ethylene glycol, Sodium iodate))

Synonym(s):

Cytology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco® (For Microscopy (contains ethylene glycol, Sodium iodate))

mp

16.67 °C

density

1.13 g/cm3

λmax

545-560 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

HARLECO® Nuclear staining

Application

HARLECO® Nuclear staining
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Liq. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service