Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65043-M

Sigma-Aldrich

Wright Stain Pack

HARLECO® Designed for use with the Ames Hema-Tek Stainer

Synonym(s):

Hematology Stain Hematrek, Hematology Solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

transition temp

flash point 15.8 °C

λmax

520-530 nm (Eosin)
645-655 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

Application

HARLECO® Designed for use with the Ames Hema-Tek Stainer
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Designed for use with the Ames Hema-Tek Stainer.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service