647950-M
Triclosan
A potent antimicrobial agent that inhibits bacterial fatty acid synthesis by selectively targeting Fabl encoded enoyl-acyl carrier protein (ACP) reductase.
Synonym(s):
Triclosan, 5-Chloro-2-(2,4-dichlorophenoxy)phenol
About This Item
Recommended Products
Quality Level
assay
≥97% (HPLC)
form
crystalline powder
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
ethanol: 50 mg/mL
storage temp.
−20°C
InChI
1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H
InChI key
XEFQLINVKFYRCS-UHFFFAOYSA-N
General description
Warning
Reconstitution
Other Notes
Surolia, N. and Surolia, A., 2001. Nat. Medicine7, 167.
Heath, R.J., et al. 2000. J. Biol. Chem.275, 4654.
Heath, R.J., et al. 1999. J. Biol. Chem.274, 11110.
Hoang, T.T. and Schweizer, H.P. 1999. J. Bacteriol.181, 5489.
Heath, R.J., et al. 1998. J. Biol. Chem. 273, 30316.
McMurray, L.M., et al. 1998. Nature394, 531.
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service