Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64000SM

Sigma-Aldrich

Midas® III-Plus Service Manual

for use with Midas III-Plus Automated Stainer

Synonym(s):

Stainer

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$580.00

$580.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
1 PKG
$580.00

About This Item

UNSPSC Code:
41105202

$580.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

compatibility

for use with Midas III-Plus Automated Stainer

shipped in

ambient

General description

For the MIDAS® III-Plus automated slide stainer

Application

For Midas and Mirastainer II Stainers. Includes Theory of Operation, Electrical Schematics, and pictures of various sub-assemblies.
MIDAS®

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service