Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616463

Sigma-Aldrich

TGF-β RI Kinase Inhibitor IX

Synonym(s):

TGF-β RI Kinase Inhibitor IX, 4-((4-((2,6-Dimethylpyridin-3-yl)oxy)pyridin-2-yl)amino)benzenesulfonamide, ALK5 Inhibitor IX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N4O3S
Molecular Weight:
370.43
UNSPSC Code:
12352200
NACRES:
NA.28

assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable pyridinyloxypyridine compound that acts as a potent and highly selective inhibitor against TGFβ RI/ALK5 (IC50 = 72 nM, [ATP] = 1 mM) by targeting both the ATP-binding site and the “selectivity pocket”, while exhibiting much reduced or little activity against a panel of 75 other kinases (IC50<1 µM). Shown to reduce TGFβ1-induced nuclear translocation of a GFP-Smad2 fusion reporter in MDA-MB-468 cells (IC50 = 22 nM) in vitro and exhibit excellent oral bioavailability (F = 75%, AUC = 4.95 µM.h) with low clearance (12 ml/min/kg) in rats in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstiution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Goldberg, F.W., et al. 2009. J. Med. Chem.52, 7901.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service