Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616450

Sigma-Aldrich

TGF-β1, Human Platelets

Synonym(s):

hTGF-β1, Transforming Growth Factor-β1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤1 EU/μg Endotoxin (EU/ug of TGF-β1)

shipped in

ambient

storage temp.

−20°C

General description

Native transforming growth factor-β1 from human platelets. Promotes apoptosis in resting human B lymphocytes, glioma cells, and trigeminal neurinomal cells.
Native transforming growth factor-β1 from human platelets. Promotes apoptosis in resting human B lymphocytes, glioma cells, and trigerminal neurinomal cells.

Biochem/physiol Actions

ED₅₀ = 40-10 pg/ml as measured by the inhibition of IL-4-dependant proliferation of mouse HT-2 cells using resazurin dye as an indicator of cell proliferation.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a sterile-filtered solution of 50 µg BSA/µg TGF-β1, 35% acetonitrile, 0.1% TFA.

Preparation Note

Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, store in the refrigerator (4°C) for short-term storage or freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 months at 4°C or up to 3 months at -20°C..
Reconstitute to a concentration ≥10 µg/ml in sterile 4 mM HCl containing ≥0.1% HSA or BSA.

Other Notes

Lomo, J., et al. 1995. J. Immunol.154, 1634.
Marushige, K. and Marushige, Y. 1995. Anticancer Res.14, 2419.
Bermundez, L.E. 1993. J. Immunol.150, 1838.
Hamilton, J.A., et al. 1993. J. Immunol.150, 1496.
Reilly, J.T., et al. 1993. Br. J. Haematol.83, 58.
Sporn, M.B. and Roberts, A.B. 1993. Growth Factors8, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Zhao et al.
Molecular cancer therapeutics, 11(2), 503-513 (2011-12-14)
A growing body of data indicates that inhibiting the type 1 insulin-like growth factor receptor (IGF-1R) might be an effective treatment strategy for hepatocellular carcinoma (HCC). OSI-906 is a dual IGF-1R/IR kinase inhibitor currently in phase II clinical development for
Uyen Q Le et al.
Scientific reports, 12(1), 19504-19504 (2022-11-15)
Triple negative breast cancer (TNBC) is a disease of poor prognosis, with the majority classified as the basal-like subtype associated with epithelial-mesenchymal transition and metastasis. Because basal breast cancers originate from proliferative luminal progenitor-like cells upon dysregulation of proper luminal
Gretchen M Argast et al.
Cells, tissues, organs, 193(1-2), 114-132 (2010-11-03)
Epithelial to mesenchymal transition (EMT) plays a dual role in tumor progression. It enhances metastasis of tumor cells by increasing invasive capacity and promoting survival, and it decreases tumor cell sensitivity to epithelial cell-targeting agents such as epithelial growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service