Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

612084

Sigma-Aldrich

TIMP-2, Human Rheumatoid Synovial Fibroblast

Native TIMP-2 from human rheumatoid synovial fibroblast.

Synonym(s):

Tissue Inhibitor of Metalloproteinase-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

Storage temp.

−70°C

General description

Native TIMP-2 from human rheumatoid synovial fibroblast. Member of a family of inhibitors that participate in the activation and regulation of MMP activity. Forms a non-covalent stoichiometric complex with latent and active MMPs. Shown to inhibit the activities of MMP-1, MMP-2, MMP-12, and transin.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP-1
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% Brij® 35 Detergent, pH 7.0.

Preparation Note

Prepared from fibroblasts that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

Greene, J., et al. 1996. J. Biol. Chem. 271, 30375.
Miyazaki, K., et al. 1993. J. Biol. Chem. 268, 14387.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service