Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

575547

Sigma-Aldrich

Tankyrase1/2 Inhibitor IV, JW55

The Tankyrase1/2 Inhibitor IV, JW55 controls the biological activity of Tankyrase1/2. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

Tankyrase1/2 Inhibitor IV, JW55, TNKS1/2 Inhibitor IV, Wnt Pathway Inhibitor XVII, N-(4-((4-(4-Methoxyphenyl)tetrahydro-2H-pyran-4-yl)methylcarbamoyl)phenyl)furan-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$214.00

$214.00


Check Cart for Availability


Select a Size

Change View
25 MG
$214.00

About This Item

Empirical Formula (Hill Notation):
C25H26N2O5
CAS Number:
Molecular Weight:
434.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$214.00


Check Cart for Availability

Quality Level

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NCC1(CCOCC1)C2=CC=C(C=C2)OC)C3=CC=C(NC(C4=CC=CO4)=O)C=C3

InChI

1S/C25H26N2O5/c1-30-21-10-6-19(7-11-21)25(12-15-31-16-13-25)17-26-23(28)18-4-8-20(9-5-18)27-24(29)22-3-2-14-32-22/h2-11,14H,12-13,15-17H2,1H3,(H,26,28)(H,27,29)

InChI key

ZJZWZIXSGNFWQQ-UHFFFAOYSA-N

General description

A cell-permeable phenylcarbamoylfuran-carboxamide compound that acts as a potent, selective, and reversible inhibitor of poly(ADP-ribosyl)ation activity of tankyrase 1 and 2 (IC50 = 1.9 and 0.83 µM, respectively). Exhibits high selectivity over PARP1 (IC50 = 20 µM). Stabilizes and increases AXIN2 levels, thereby enhancing phosphorylation and degradation of β-catenin resulting in reduced canonical Wnt signaling (IC50 = 470 nM in Wnt 3a-induced HEK293 cells transfected with ST-Luc). Shown to induce cell cycle arrest and diminish the proliferation of SW480 colorectal cancer cells (~ 5 µM). Suppresses tumor growth in tamoxifen-induced polyposis in Apc knock-out murine model (100 mg/kg, i.p.). Displays adequate human liver microsomal stability (t1/2 = 10.1 min).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
tankyrase 1 and 2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Waaler, J., et al. 2012. Cancer Res.72, 2822.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service