574712
Syk Inhibitor II
The Syk Inhibitor II, also referenced under CAS 227449-73-2, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Syk Inhibitor II, 2-(2-Aminoethylamino)-4-(3-trifluoromethylanilino)-pyrimidine-5-carboxamide, Dihydrochloride, Dihydrate
About This Item
Recommended Products
Quality Level
assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
off-white
solubility
DMSO: 100 mg/mL
water: 25 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C14H15F3N6O.2ClH.2H2O/c15-14(16,17)8-2-1-3-9(6-8)22-12-10(11(19)24)7-21-13(23-12)20-5-4-18;;;;/h1-3,6-7H,4-5,18H2,(H2,19,24)(H2,20,21,22,23);2*1H;2*1H2
InChI key
LCACSMNPIITJGG-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Syk
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service