Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

569615-M

Sigma-Aldrich

Stem-Cell Factor/c-Kit Inhibitor, ISCK03 - CAS 945526-43-2 - Calbiochem

Synonym(s):

4-t-Butylphenyl-N-(4-imidazol-1-yl-phenyl)sulfonamide, SCF/c-Kit Signaling Inhibitor, ISCK03, c-Kit Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥97% (HPLC)

form

solid

color

dark yellow

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

General description

The Stem-Cell Factor/c-Kit Inhibitor, ISCK03, controls the biological activity of Stem-Cell Factor/c-Kit. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Joo Na et al.
Biochemical pharmacology, 74(5), 780-786 (2007-07-31)
It is well known that c-kit is related to pigmentation as well as to the oncology target protein. The objective of this study was to discover a skin-whitening agent that regulates c-kit activity. We have developed a high-throughput screening system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service