Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

569372

Sigma-Aldrich

PhosphoDetect Anti-Src (pTyr⁴¹⁸) Mouse mAb (9A6)

lyophilized, clone 9A6, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9A6, monoclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

Storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr418)

General description

Purified mouse monoclonal antibody. Recognizes the ~60 kDa Src protein phosphorylated at Tyr418 (human, rat), Tyr423 (mouse), and Tyr416 (chicken, v-Src).
Recognizes the ~60 kDa Src protein phosphorylated at Tyr418 in chick embryo fibroblasts expressing human wild-type Src.
This PhosphoDetect Anti-Src (pTyr⁴¹⁸) Mouse mAb (9A6) is validated for use in ELISA, Immunoblotting, Immunocytochemistry for the detection of Src (pTyr⁴¹⁸).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the pTyr⁴¹⁸ phosphorylation site of human Src

Application

ELISA (0.05 µg/ml)

Immunoblotting (1 µg/ml, colorimetric; 0.1 µg/ml, chemiluminescence)

Immunocytochemistry (1 µg/ml, immunofluorescence)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 2X PBS, PEG, sucrose.

Reconstitution

Reconstitute in 1 ml sterile dH₂O (15 min, room temperature). Following reconstitution, refrigerate (4°C) for short-term storage or aliquot, flash-freeze in liquid nitrogen, and freeze (-70°C) for long-term storage. Stock solutions are stable for up to 3 months at 4°C or for up to 1 year at -70°C. Thaw aliquots at 37°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Does not cross-react with non-phosphorylated Src, or with unrelated phosphorylation sites. The sequence of the immunogen is conserved in human (Tyr418), mouse (Tyr423), rat (Tyr418), chicken (Tyr416), and c-Src (Tyr416). Variables associated with assay conditions will dictate the optimal working dilution.
Martin, G.S. 2001. Nat. Rev. Mol. Cell Biol.2, 467.
Reissig, D., et al. 2001. J. Cancer Res. Clin. Oncol.127, 226.
Bjorge, J.D., et al. 2000. Oncogene19, 5620.
Irby, R.B. and Yeatman, T.J. 2001. Oncogene19, 5636.
Miermont, A.M., et al. 2000. J. Immunol. Methods246, 203.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service