Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565791

Sigma-Aldrich

γ-Secretase Inhibitor XXII , Aβ42-Selective, CS-1

The γ-Secretase Inhibitor XXII, Aβ42-Selective, CS-1, also referenced under CAS 1210283-33-2, controls the biological activity of γ-Secretase.

Synonym(s):

γ-Secretase Inhibitor XXII , Aβ42-Selective, CS-1, 3,3ʹ-(3,4,5-Trifluorophenyl)methylenebis-(4-hydroxycoumarin)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H13F3O6
CAS Number:
Molecular Weight:
466.36
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable di-coumarin compound that selectively inhibits γ-secretase-mediated production of Aβ42 over Aβ38, Aβ40, and Notch1 both in cell-free APP cleavage assays (IC50 = 0.07, 0.71, 0.31, and 1.77 µM, respectively) and in human and murine cultures (100% blockage of Aβ42 production and no effect against the rest at 30 µM) by altering the S1′ and S3′ pockets within the presenilin substrate binding site via an allosteric, non-substrate-competitive mechanism.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shelton, C.C., et al. 2009. Proc. Natl. Acad. Sci. USA106, 20228.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service