Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559240

Sigma-Aldrich

Anti-rSec6 Mouse mAb (9H5)

liquid, clone 9H5, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

9H5, monoclonal

form

liquid

does not contain

preservative

species reactivity

chicken, rabbit, bovine, human, monkey, mouse, rat, canine, porcine, hamster

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Immunoaffinity purified mouse monoclonal antibody. Recognizes the ~87 kDa rSec6 protein.
Recognizes the ~87 kDa rSec6 protein in PC12 cells.
This Anti-rSec6 Mouse mAb (9H5) is validated for use in Immunoblotting, Immunofluorescence for the detection of rSec6.

Immunogen

Rat
recombinant, rat rSec6 fused to a His•Tag sequence

Application

Immunoblotting (4 µg/ml, chemiluminescence)

Immunofluorescence (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.1 mM PMSF, 50% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
ESK-4 cells

Other Notes

Immunofluorescence of PC12 cells using this antibody shows colocalization of rSec6 with vesicles and sites of exocytosis. Variables associated with assay conditions will dictate the proper working dilution.
Kee, Y., et al. 1997. Proc. Natl. Acad. Sci. USA94, 14438.
Hsu, S.-C., et al. 1996. Neuron17, 1209.
TerBush, D.R., et al. 1995. J. Cell Biol.130, 299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Wersäll et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(4), 787-800 (2018-02-14)
RalA and RalB GTPases are important regulators of cell growth, cancer metastasis, and granule secretion. The purpose of this study was to determine the role of Ral GTPases in platelets with the use of platelet-specific gene-knockout mouse models. This study
Annie Morin et al.
Small GTPases, 1(3), 142-156 (2011-06-21)
RhoGDIs are negative regulators of small GTP-binding proteins of the Rho family, which have essential cellular functions in most aspects of actin-based morphology and motility processes. They extract Rho proteins from membranes, keep them in inactive rhoGDI/Rho complexes and eventually

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service