Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5450-OP

Sigma-Aldrich

L-Histidine

OmniPur® Grade, dried basis: 98.5-101.5%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solid

manufacturer/tradename

Calbiochem®

color

white

shipped in

ambient

General description

Molecular biology grade.

Specifications:
Assay (dried basis): 98.5-101.5%
Chloride (Cl): ≤0.05%
DNase: None detected
Heavy metals (as Pb): ≤0.0015%
Identification (IR-spectrum): Conforms
Residue after ignition: ≤0.4%

Iron (Fe): ≤0.003%
Loss on drying (105°C): ≤0.2%
Protease: None detected
RNase: None detected
Specific rotation [a]25/D: +12.4-+14.0°
Sulfate (SO4): ≤0.03%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service