Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539732-M

Sigma-Aldrich

PTP, T-Cell, Human, Recombinant, E. coli

Synonym(s):

Protein Tyrosine Phosphatase, T-Cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥10,000 units/mL
≥15,000 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

Recombinant, human T-cell PTP, truncated at the C-terminus (an 11Kd deletion) and expressed in E. coli. Suitable for dephosphorylation of phosphotyrosine residues in proteins.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 nmol of pNPP per min at 30°C, pH 7.0.

Physical form

In 100 mM NaCl, 50 mM HEPES, 5 mM DTT, 2 mM EDTA, 0.01% Brij® 35 Detergent, 50% glycerol, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Gordon, J.A. 1991. Methods Enzymol.201, 477.
Zander, N.F., et al. 1991. Biochemistry30, 6964.
Cool, D.E., et al. 1989. Proc. Natl. Acad. Sci. USA86, 5257.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service