Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38768

Sigma-Aldrich

Lim Kinase Inhibitor II, SR-7826

Synonym(s):

Lim Kinase Inhibitor II, SR-7826, 1-(2-Hydroxyethyl)-3-(4-(5-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)phenyl)-1-phenylurea, LIMK Inhibitor, SR-7826

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$173.00

$173.00


Check Cart for Availability


Select a Size

Change View
5 MG
$173.00

About This Item

Empirical Formula (Hill Notation):
C22H21N5O2
Molecular Weight:
387.43
UNSPSC Code:
12352200
NACRES:
NA.77

$173.00


Check Cart for Availability

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell permeable bis-aryl urea that acts as a highly potent and selective LIMK inhibitor (LIM kinase, IC50 = 43 nM for Limk1) with more than 400-fold selectivity against ROCK and JNK kinases. Only displays inhibitory effect on STK16 with ≥80% inhibition at 1 µM in addition to LIMK1 in a 61 kinase panel profiling. Inhibits cofilin phosphorylation in A7r5 (IC50 = 470 nM), PC-3 and CEM-SS T cells (IC50<1 µM), shown to inhibit cell invasion by 76% and cell migration by 74% in PC-3 cells at 1 µM. Exhibits desirable pharmacokinetic properties in vitro and in vivo with 36% bioavailability in rats (p.o. at 2mg/kg).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LIMK
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C

Other Notes

Yin, Y., et al. 2015. J. Med. Chem.58, 1846.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service