Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.38384

Sigma-Aldrich

NCK/PAK Blocking Peptide

Synonym(s):

NCK/PAK Blocking Peptide, TAT-PAK peptide, NCK blocking peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C126H221N51O32S2 · 11C2HF3O2 · 7H2O
Molecular Weight:
4406.92
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable synthetic peptide derived from the proline-rich region near the N-terminal of p21 activated kinase1 (PAK1) and fused with HIV TAT. Shown to selectively binds to the non-catalytic region of tyrosine kinase adaptor protein (NCK) and disrupts the cellular transport and localization of PAK. Shown to specifically inhibit endothelial cell migration and contractility (IC50 = 2 µM) and block bFGF-induced angiogenesis in a chick chorioallantoic membrane (CAM) assay. Does not exhibit any deleterious effects on endothelial cell survival and growth.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NCK
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H2N-YGRKKRRQRRRGKPPAPPMRNTSTM-C02H

Physical form

Supplied as TFA salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Chen, J., et al. 2015, Sci. Signal.8, 365.

Kiosses, W., et al. 2002, Circ. Res.90, 697.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service