Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537012-M

Millipore

Prolyl Endopeptidase Inhibitor III, KYP-2047

The Prolyl Endopeptidase Inhibitor III, KYP-2047 controls the biological activity of Prolyl Endopeptidase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Prolyl Endopeptidase Inhibitor III, KYP-2047, (S)-1-((S)-1-(4-phenylbutanoyl)-pyrrolidine-2-carbonyl)pyrrolidine-2-carbonitrile, 4-phenylbutanoyl-Lprolyl-2(S)-cyanopyrrolidine, PREP inhibitor III, KYP-2047

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N3O2
Molecular Weight:
339.43
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥98% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear colorless

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, potent, fast-acting, and selective inhibitor of prolyl oligopeptidase (PREP) (Ki = 23 pM). Reported to cross the blood-brain barrier. Inhibits about 85% PREP activity within 10 minutes of administration and exhibits a half-life of about 4.8 hours. Exhibits higher lipophilicity that increases its penetration across biological membranes in vivo. Maximum plasma and brain levels are achieved within 10 minutes following its i.p. administration. Shown to block oxidative-stress induced α-synuclein aggregation and clearance in cellular and animal models of Parkinson′s disease. Also shown to improve cognitive performance in young rats (1-5 mg/kg) in a dose-dependent manner.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jarho, E., et al. 2004. J. Med. Chem.47, 5605.
Venalainen, J., et al. 2011. Biochem. Pharmacol.71, 683.
Jalkanen, A., et al. 2006. Basic & Clin. Pharmac. & Tox.100, 132.
Jalkanen, A., et al. 2011. Basic & Clin. Pharmac. & Tox.109, 443.
Klimaviciusa, L., et al. 2012. Journ. Neuros. Methods.204, 104.
Myohanen, T., et al. 2012. Brit. J. Pharm.166, 1097.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service