Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.34330

Sigma-Aldrich

MALT1 Inhibitor III, Mepazine Acetate

Synonym(s):

MALT1 Inhibitor III, Mepazine Acetate, Pacatal, 10-((1-Methyl-3-piperidinyl)methyl)-10H-phenothiazine, Acetate, Mucosa-Associated Lymphoid tissue Lymphoma Translocation 1 Inhibitor III, MALT-1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2S · xC2H4O2
Molecular Weight:
310.46 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Product 5.34330 is not currently sold in your country. Contact Technical Service

assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

purple

solubility

DMSO: 100 mg/mL
water: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable phenothiazine that inhibits MALT1, but not caspase-3 or -8, proteolytic activity (IC50 = 0.83 and 0.42 µM, respectively, against full-length or paracaspase domain-containing aa 325-760 MALT1 GST fusion-catalyzed Ac-LRSR-AMC hydrolysis) in a reversible and noncompetitive manner and blocks anti-CD3/CD28-stimulated IL-6 production (20 h) in primary murine CD4+ T-cell (by 77%) and human PBMC cultures (by 51 - 89%). Shown to suppress ABC-DLBCL constitutive cellular MALT1 activity (>75% suppression by 10 µM Mepazine; IC50<5 µM in U2932, OCI-Ly10, OCI-Ly3, HBL1, and TMD8 cultures) and RelB cleavage, resulting in effective blockage of NF-κB-dependent IL-6, IL-10, as well as anti-apoptotic Bcl-xL and FLIP-L expressions. Mepazine exhibits ABC-DLBCL-selective cytotoxicity (≥27% cell death in 4 days by 10 µM Mepazine in HBL-1, OCI-Ly3, OCI-Ly10, TMD8, and U2932 cultures) over GCB-DLBCL (≤26% cell death in 4 days by 20 µM Mepazine in DJAB, Su-DHL-4, and Su-DHL-6 cultures) both in cultures in vitro and in mice (74%, and 0% suppression, respectively, of OCI-Ly10 and Su-DHL-6 tumor expansion on day 22 post cancer transplant; via daily 400 µg/kg i.p.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MALT1
Reversible: yes
Target IC50: 0.83 and 0.42 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schlauderer, F., et al. 2013. Angew. Chem. Int. Ed.52, 10384.
Nagel, D., et al. 2012. Cancer Cell.22, 825.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service