Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33641

Sigma-Aldrich

CHOP Activator, Cpd12

Synonym(s):

CHOP Activator, Cpd12, N-(4-((4-chloropiperidin-1-yl)sulfonyl)phenyl)-5-nitrofuran-2-carboxamide, C/EBP-Homologous Protein Activator, Cpd12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O6S
CAS Number:
Molecular Weight:
413.83
UNSPSC Code:
12352200

assay

≥96% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

Storage temp.

2-8°C

General description

A cell-permeable sulfonamidebenzamide derivative that acts as a potent and selective inducer of C/EBP-homologous protein (CHOP; AC50 = 800 nM) without affecting XBP (AC50 >80 µM). Displays a balanced profile in terms of its lipophilic profile (cLogP = 2.7), selectivity, and activation of CHO-CHOP luciferase reporter system. Exhibits high selectivity against a panel of 67 GPCR, nuclear receptors, transporters, and ion channels. Has significantly reduced activity towards dopamine receptor at high concentrations (68% at 10 µM). upregulates the expression of UPR target genes. Displays anti-proliferation properties in wild-type mouse embryonic fibroblasts (EC50 = 4.8 µM), but does not affect CHOP knock out cells even at high concentrations (EC50 >20 µM). Inhibits the proliferation of several cancer cell lines at low micromolar levels. Exhibits favorable pharmacokinetic profile (Cmax = 140 nM; Tmax = 0.17 h at 30 mg/kg, i.p. single dose).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CHOP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Flaherty, D. P., et al. 2014. ACS Med Chem Lett.5, 1278.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service