Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33313

Sigma-Aldrich

Pyruvate Kinase Inhibitor III - CAS 313701-92-7 - Calbiochem

Synonym(s):

Pyruvate Kinase Inhibitor III - CAS 313701-92-7 - Calbiochem, 5-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-hydroxybenzoic acid, PKM2 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO3
CAS Number:
Molecular Weight:
231.25
UNSPSC Code:
12352200

assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown to brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C13H13NO3/c1-8-3-4-9(2)14(8)10-5-6-12(15)11(7-10)13(16)17/h3-7,15H,1-2H3,(H,16,17)

InChI key

LBDOGIZEEBHJSE-UHFFFAOYSA-N

General description

A cell-permeable pyrrolyl-hydroxybenzoate compound that inhibits fructose-1,6-bisphosphate- (FBP-) dependent activation of pyruvate kinases PKM2 and PKL (IC50 10 to 50 µM), while exhibiting reduced potency against non-FBP-dependent PKM1 activity (81% inhibition of PKM2 vs. 34% inhibition of PKM1 at 30 µM). Lysates from inhibitor-treated (250 µM inhibitor treatment for 6 h) human lung cancer H1299 cells are shown to exhibit significantly reduced PKM2 activity (by 84%) even upon exogenous FBP addition. Inhibitor treatment (Effective conc. 50 to 100 µM) is also demonstrated to enhance cell death of IL-3-dependent murine pro-B lymphocytic FL5.12 and EGFR-signaling-dependent human lung adenocarcinoma HCC827 cultures upon IL-3 withdrawal and EGFR inhibitor treatment, respectively.
A cell-permeable pyrrolyl-hydroxybenzoate compound that inhibits fructose-1,6-bisphosphate- (FBP-) dependent activation of pyruvate kinases PKM2 and PKL (IC50 10 to 50 µM), while exhibiting reduced potency against non-FBP-dependent PKM1 activity (81% inhibition of PKM2 vs. 34% inhibition of PKM1 at 30 µM). Lysates from inhibitor-treated (250 µM inhibitor treatment for 6 h) human lung cancer H1299 cells are shown to exhibit significantly reduced PKM2 activity (by 84%) even upon exogenous FBP addition. Inhibitor treatment (Effective conc. 50 to 100 µM) is also demonstrated to enhance cell death of IL-3-dependent murine pro-B lymphocytic FL5.12 and EGFR-signaling-dependent human lung adenocarcinoma HCC827 cultures upon IL-3 withdrawal and EGFR inhibitor treatment, respectively.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
pyruvate kinases
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Vander Heiden, M.G., et al. 2010. Biochem. Pharmacol.79, 1118.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service