Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.33196

Sigma-Aldrich

Myogenic miRNA Inhibitor, Cpd1

Synonym(s):

Myogenic miRNA Inhibitor, Cpd1, tert-Butyl (5-(8a-methoxy-3,8-dioxo-2-phenyl-2a,3,8,8a-tetrahydrocyclobuta[b]naphthalen-1-yl)quinolin-8-yl) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H27NO6
CAS Number:
Molecular Weight:
533.57
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

potency

2.5 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrahydroquinolinedione[b]cyclobutene based compound that acts as a specific inhibitor of microRNA (miRNA)-221/222-myoD regulatory pathway. Inhibits the translation of myoD in C2C12 cells without changing the expression level of myoD mRNA, but reduces myoD protein expression levels. Shown to selectively inhibit miR-1 (IC50 = 2.5 µM), miR-133a and miR-206 (IC50 = 5 µM) in C2C12 cells. However, it does not affect the levels of non-myogenic miR-9, miR-25, miR-150, miR-25, and miR-214. Shown to inhibit the differentiation of C2C12 cells as indicated by the down-regulation of muscle creatine kinase, myosin heavy chain, and myoGenin levels in these cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
miR-221/22

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tan, S., et al. 2014. Chem. & Biol.21, 1265.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service