5.33003
Ammonia solution 25%
for LC-MS LiChropur™
Synonym(s):
Ammonium hydroxide solution, Ammonium hydroxide solution, Ammonia water, Ammonia aqueous, Ammonia water
Select a Size
$113.00
Select a Size
About This Item
$113.00
Recommended Products
vapor pressure
483 hPa ( 20 °C)
Quality Level
expl. lim.
27 %
technique(s)
HPLC: suitable
bp
37.7 °C/1013 hPa
mp
-57.5 °C
density
0.9 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
[NH4+].[OH-]
InChI
1S/H3N.H2O/h1H3;1H2
InChI key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
related product
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
8B - Non-combustible, corrosive hazardous materials
wgk_germany
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn the tips and tricks of selecting and preparing a suitable mobile phase buffer for your HPLC separations and method developments for samples that contain ionizable compounds.
Protocols
We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service