Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.32585

Sigma-Aldrich

Fatty Acid Synthase Inhibitor II, GSK2194069

Synonym(s):

Fatty Acid Synthase Inhibitor II, GSK2194069, FAS Inhibitor II, ( S)-4-(4-(Benzofuran-5-yl)phenyl)-3-((1-(cyclopropanecarbonyl)pyrrolidin-3-yl)methyl)-1H-1,2,4-triazol-5(4H)-one, FASN Inhibitor II, GSK 2194069

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N4O3
CAS Number:
Molecular Weight:
428.48
MDL number:
UNSPSC Code:
12352200

assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C25H24N4O3/c30-24(18-1-2-18)28-11-9-16(15-28)13-23-26-27-25(31)29(23)21-6-3-17(4-7-21)19-5-8-22-20(14-19)10-12-32-22/h3-8,10,12,14,16,18H,1-2,9,11,13,15H2,(H,27,31)/t16-/m0/s1

InChI key

AQTPWCUIYUOEMG-INIZCTEOSA-N

General description

A cell-permeable, aqueous soluble (79 µg/mL or 184 µM) triazolone compound that selectively inhibits the β-ketoacyl reductase (KR) activity of fatty acid synthase (FAS) in a reversible (IC50 in nM/h of pre-incubation = 13/0, 16/1, 15/4.7, 14/7; 2 nM hFAS/4 mM trans-1-decalone/ 48 nM drug), trans-1-decalone-competitive (Ki = 10 nM), and NADPH-uncompetitive manner by targeting FAS in its NADPH-bound, but not free or NADP+-bound form, while exhibiting little or no potency toward FAS β-ketoacyl synthase (KS), β-hydroxyoacyl dehydratase (DH), enoyl reductase (ER), or thioesterase (TE) domain activity. Shown to effectively inhibit de novo FA synthesis in cancer cultures (by >70% in KATO-III, MKN45, SNU-1, and A549 cells;100 nM for 24 h) with a concomitant accumulation of cellular FAS substrate malonyl-CoA (13-fold of control using A549 cells; 30 nM for 48 h) and a drop in phosphatidylcholine level (IC50 = 15.5 nM in 48 h-treated A549 cultures), resulting in an effective growth inhibition in A549 cultures (GI50/5 d = 15 nM) reversible with palmitate co-treatment.
GSK2194069; 1332331-08-4; FASN Inhibitor; FAS Inhibitor

Biochem/physiol Actions

Cell permeable: yes
Primary Target
β-ketoacyl reductase (KR) activity of fatty acid synthase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hardwicke, M.A., et al. 2014. Nat. Chem. Biol.10, 774.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service