Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.32129

Millipore

Intracellular pH Sensor, TPE-Cy

Synonym(s):

pHi Sensor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-orange

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable TPE-Cy adduct that acts as a two-channel ratiometric fluorogen and is sensitive to intracellular pH in the entire physiological range.
A cell-permeable, biocompatible tetraphenylethene-cyanine (TPE-Cy) adduct that acts as a two-channel ratiometric fluorogen and is sensitive to intracellular pH in the entire physiological range. Displays a large Stokes shift and exhibits aggregation-induced emission (AIE). Following its entry into live cells, it is distributed in the cytoplasm, accumulates in organelles, and becomes fluorescent. Interacts with the lipid components of the cells and shifts its color transition point of the red-to-blue emission from extracellular pH 10 to intracellular physiological pH range. At neutral pH it provides a strong blue emission (peak at 489 nm), which is enhanced at pH 8.07. However, acidification of medium causes a reduction in the blue emission (489 nm) and induces red emission (peak at 615 nm). The ratiometric red and blue signals can serve as an indicator for local proton concentration in the cell. Does not affect cell viability up to 12.5 µM concentration.

Biochem/physiol Actions

Cell permeable: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, S., et al. 2013. J. Am. Chem. Soc.135, 4926.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service