Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.31709

Sigma-Aldrich

Apoptosis Activator IX, DPP-23

Synonym(s):

Apoptosis Activator IX, DPP-23, Unfolded Protein Response Inducer, DPP-23, UPR Inducer, DPP_23

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable chalcone derivative that enhances the production of reactive oxygen species and reduces glutathione levels selectively in cancer cells while sparing normal cells and induces unfolded protein response (UPR) through up-regulating UPR related genes. Inhibits the growth of in multiple cancer cell lines (Capan-1, MDA-MB231, HCT116, and HT1080 cells) and suppresses the growth of HCT116 tumor xenografts in nude mice via the induction of apoptosis (10 mg/kg, i.p; q.d.). Arrests cell cycle at the G2/M phase and triggers protective autophagy and induces apoptosis by increasing the active forms of caspase 3, 7, and 9. Treatment of serum-starved MIA PaCa-2 cells with DPP-23 induces the phosphorylation of ERK1/2, JNK1/2, and p38 MAPK in a time-dependent manner.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shin, Y.S., et al. 2014. Clin. Cancer Res.20, in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service