Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

5.31621

Sigma-Aldrich

APT2 Inhibitor, Cpd1

Synonym(s):

APT2 Inhibitor, Cpd1, Lysophosphilipase 2 Inhibitor, LYPLA2 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22N2O4S2
CAS Number:
Molecular Weight:
454.56
MDL number:
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

potency

510 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale orange

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=C(C(C=CC=C3)=C3S(C2)(=O)=O)S1)N(CC4)CCN4C5=CC=C(OC)C=C5

General description

A cell permeable acetylpiperazine derivative that acts as a selective and reversible inhibitor of lysophopholipase 2 (LYPLA2; IC50 = 510 nM, Ki = 230 nM) without affecting the activity of LYPLA1 and 25 other serine hydrolases in mouse BW5147 T cell hydridoma proteome. Shown to cause an almost complete inhibition (+90%) of LYPLA2 activity in lung, heart, and kidney (50 mg/kg, 3 h), and in HEK293T, and mouse T cells (~ 5 µM for 3 h).
A cell permeable, selective, and reversisble inhibitor of lysophopholipase 2 (LYPLA2; IC₅₀ = 510 nM, Ki = 230 nM). Does not affect LYPLA1.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
APT2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Adibekian, A., et al. 2012. J. Am. Chem. Soc.134, 10345.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service