Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30820

Sigma-Aldrich

Rac1 Inhibitor V

Synonym(s):

Rac1 Inhibitor V, 3-(2-Hydroxyphenyl)-N-(4-(1-piperidinylsulfonyl)phenyl)-1H-pyrazole-5-carboxamide, TCMDC-124310

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N4O4S
CAS Number:
Molecular Weight:
426.49
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)CNC1=NC(=NC=C1)NCC2=CSC(=N2)C3=CC=CC=C3

General description

A cell-permeable {piperidinylsulfonylphenyl}-pyrazolocarboxamide compound that inhibits celluar Rac-GTP level upon PDGF-BB stimulation (Cat. No. 521225; 20 ng/mL for 2 min) in 48 h 0.4% FCS-starved A617 human vascular smooth muscle cell (SMC) cultures with a higher potency (IC50 = 8.7 µM; 4 h drug preincubation) than Z62954982 (Cat. Nos. 553511 & 553512; 37% inhibition at 50 µM) via a similar mode of action as NSC23766 (Cat. Nos. 553502 & 553508), exhibiting no potency against cellular RhoA-GTP level even at a high concentration of 25 µM. Effectively inhibits SMC adhesion (IC50 = 8.7 µM post 4 h drug treatment) and PDGF-BB-stimulated lamellipodia formation (complete inhibition with 4 h 10 µM; drug treatment prior to 10 min 10 ng/mL PDGF-BB stimulation) of 48 h starved A617 cells. Effectively inhibits Rac activation mediated by Rac-specific Tiam-1 & TrioN GEF as well as non-Rac-specific GEF Vav2, while EHop-016 (Cat. No. 553513), in comparison, is selective against Vav2- over Tiam-1-mediated Rac activation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rac
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ferri, N., et al. 2013. Med. Chem. Commun.4, 537.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service