Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30266

Sigma-Aldrich

Cytohesin Inhibitor II, Cyplecksin 2

Synonym(s):

Cytohesin Inhibitor II, Cyplecksin 2, 5-Benzyl-5-bromo-1-(4-chlorophenyl)pyrimidine-2,4,6(1 H,3 H,5 H)-trione, Cytohesin pleckstrin homology domain inhibitor 2, Cyth PH Domain Inhibitor, 5-Benzyl-5-bromo-1-(4-chlorophenyl)pyrimidine-2,4,6(1H,3H,5H)-trione, Cytohesin pleckstrin homology domain inhibitor 2, Cyth PH Domain Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12BrClN2O3
CAS Number:
Molecular Weight:
407.65
UNSPSC Code:
12352200

assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

ethanol: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable 5-bromobarbiturate that selectively and covalently modifies the pleckstrin homology (PH) domain of the cytohesin family (Kd ~ 2 µM to cytohesin-1 labeled with Alexa647) and irreversibly inhibits its function. Prevents the interaction cytohesin-1 PH, and to cytohesins 1 and 2 lacking the short C-terminal polybasic region with PIP3 and PIP2. Blocks insulin-dependent cytohesin-2-GFP translocation to plasma membrane in HeLa cells (~50 µM). Does not recognize or show any binding to T-lymphoma invasive and metastasis inducing protein 1 (Tiam)-DH-PH and shows poor affinity towards Sec7 domain of cytohesin-2. Cytohesins Inhibitor SecinH3 is also available (>Cat. No. 565725).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable 5-bromobarbiturate that targets the PH domain of cytohesin family small GEFs (Kd ~2 µM in binding assay using Cyth1 PH domain aa259-381) via covalent interaction and effectively competes against both PIP2 and PIP3 for Cyth PH domain binding (IC50 ≤10 µM against 30 nM PIP2; [Cyth1 PH] = 250 nM), while exhibiting no affinity toward Tiam1 DH-PH domain and displaying little potency against GEP100-PH/PIP2, DAGK- PH/PIP2, ARHGAP25-PH/PIP3, IRS1-PH/PIP2, Pleckstrin-PH/PIP3, or Akt2/PIP3 interaction even at concentrations as high as 100 µM. Overnight pretreatment (50 µM) prior to insulin stimulation (100 nM for 10 min) is shown to greatly inhibit insulin-induced Cyth2 plasma membrane translocation by 83% in HeLa Cyth2-GFP transfectants. Greatly complements the Sec7 domain-targeting SecinH3 (Cat. No. 565725) in studying Cytohesins-mediated cellular signaling events.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
pleckstrin homology domain of the cytohesin family
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Hussein, M., et al. 2013. Angew. Chem. Int. Ed.,52, 9529.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service