Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

529505

Sigma-Aldrich

PON1 Activator, ZNPA

The PON1 Activator, ZNPA modulates the biological activity of PON1. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

PON1 Activator, ZNPA, (Z)-2,3-bis (4-nitrophenyl)-acrylonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$155.00

$155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$155.00

About This Item

Empirical Formula (Hill Notation):
C15H9N3O4
CAS Number:
Molecular Weight:
295.25
UNSPSC Code:
41106609
NACRES:
NA.77

$155.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 25 mg/mL, pale yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

N#C/C(C1=CC=C(C=C1)[N+]([O-])=O)=C\C2=CC=C(C=C2)[N+]([O-])=O

InChI

1S/C15H9N3O4/c16-10-13(12-3-7-15(8-4-12)18(21)22)9-11-1-5-14(6-2-11)17(19)20/h1-9H/b13-9+

InChI key

QPMLLJYCAKQATA-UKTHLTGXSA-N

General description

A cell-permeable stilbene derivative that serves as an aryl hydrocarbon receptor (AhR) ligand and acts as a highly specific and potent inducer of paraoxonase 1 (PON1). Shown to be a better activator of PON1 mRNA (EC50 = 1 µM) and inducer of PON1 enzymatic activity (>3 fold at 5 µM) when compared to resveratrol (Cat. No. 554325). However, it is devoid of any activity against estrogen receptors and is not shown to inhibit activity of tyrosine kinases. Also shown to antagonize the effects of 2,3,7,8-Tetrachlorodibenzo-p-Dioxin (TCDD) on cytochrome P450 1A1 (CYP1A1).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Guyot, E., et al. 2011. Biochem. Pharm. 83, 627.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service