525331
Photochromic Ion Channel Blocker, QAQ
Synonym(s):
Photochromic Ion Channel Blocker, QAQ, 2,2′-((diazene-1,2-diylbis(4,1-phenylene))bis(azanediyl))bis(N,N,N-triethyl-2-oxoethanaminium) formate, Quaternary Ammonium–Azobenzene–Quaternary Ammonium
About This Item
assay
≥95% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
brown
solubility
DMSO: 25 mg/mL
shipped in
wet ice
storage temp.
2-8°C
General description
Biochem/physiol Actions
Ion Channels
Packaging
Warning
Reconstitution
Other Notes
Banghart, M., et al. 2009. Angew Chem Int Ed Engl.48, 9097.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service