Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521215

Sigma-Aldrich

PDGF, AA Homodimer, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg PDGF-AA)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human platelet derived growth factor AA (disulfide-linked dimer of two A-chain monomers) expressed in E. coli. Amino acid sequence is the long form of mature human PDGF-A chain and is identical to that deduced from the native human nucleotide sequence except for the addition of an N-terminal methionine group.

Biochem/physiol Actions

1-3 ng/ml

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a sterile-filtered solution in 30% acetonitrile, 0.1% TFA.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long-term storage or refrigerate (4°C) for short-term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at 4°C.
Reconstitute to a concentration ≥10 µg/ml with sterile 4 mM HCl containing at least 0.1% HSA or BSA.

Other Notes

Butt, A.M., et al. 1997. J. Neurosci. Res. 48, 588.
Westermark, B., and Heldin, C.-H. 1991. Cancer Res.51, 5087.
Betsholtz, C., et al. 1986. Nature320, 695.
Raines, E.W., et al. 1985. Methods Enzymol.109, 749.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service