Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516570-M

Millipore

PPARγ Modulator, SR1664

A cell-permeable non-thiazolidinediones derivative that binds tightly to peroxisome proliferator-activated γ (PPAR-γ) receptor, but does not exhibit any transcriptional agonism.

Synonym(s):

PPARγ Modulator, SR1664, (S)-4′-((2,3-dimethyl-5-(1-(4-nitrophenyl)ethylcarbamoyl)-1H-indol-1-yl)methyl)biphenyl-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H29N3O5
Molecular Weight:
547.60
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥92% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable non-thiazolidinediones derivative that binds tightly to peroxisome proliferator-activated γ (PPAR-γ) receptor (Ki = 29 nM; IC50 = 80 nM), but does not exhibit any transcriptional agonism. Unlike TZD compounds, it does not induce weight gain or diminish hematocrit. Shown to effectively block CDK-5-mediated phosphorylation of Ser273 of PPAR-gamma in adipose tissue. Supresses hepatic glucose production, reduces plasma free fatty acid levels, and improves insulin sensitivity of adipose tissue.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Choi, J., et al. 2011, Nature477, 477.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service