Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513020

Sigma-Aldrich

PD 145065

A highly potent but non-selective endothelin receptor antagonist [IC₅₀= 4 nM for the ETA receptor (rabbit renal artery vascular smooth muscle cells); IC₅₀ = 15 nM for the ETB receptor (rat cerebellum)].

Synonym(s):

PD 145065

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H63N7O10 · 2Na
CAS Number:
Molecular Weight:
992.08
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

phosphate buffer, pH 7.5: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C52H67N7O10/c1-8-29(5)44(49(65)57-41(52(68)69)25-34-27-53-38-21-15-14-18-35(34)38)59-50(66)45(30(6)9-2)58-48(64)40(26-42(61)62)55-47(63)39(24-28(3)4)56-51(67)46(54-31(7)60)43-36-19-12-10-16-32(36)22-23-33-17-11-13-20-37(33)43/h10-21,27-30,39-41,43-46,53H,8-9,22-26H2,1-7H3,(H,54,60)(H,55,63)(H,56,67)(H,57,65)(H,58,64)(H,59,66)(H,61,62)(H,68,69)/t29-,30-,39-,40-,41-,44-,45-,46+/m0/s1

InChI key

HRAQSWKGRRUBDJ-OMUAVVNCSA-N

General description

A highly potent but non-selective endothelin receptor antagonist [IC50 = 4 nM for the ETA receptor (rabbit renal artery vascular smooth muscle cells); IC50 = 15 nM for the ETB receptor (rat cerebellum)].

Biochem/physiol Actions

Cell permeable: no
Primary Target
Endothelin receptor antagonist
Product does not compete with ATP.
Reversible: no
Target IC50: 4 nM against endothelin receptor (ETA receptor) in rabbit renal artery vascular smooth muscle cells; 15 nM for the ETB receptor (rat cerebellum)

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-D-Bhg-Leu-Asp-Ile-Ile-Trp-OH (Bhg = 5H-dibenzo[a,d]cycloheptene-5-glycine)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 4 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Han, H.K., et al. 1998. Life Sci. 63, 1599.
Allcock, G.H., et al. 1995. Br. J. Pharmacol.116, 2482.
Wellings, R.P., et al. 1994. Eur. J. Pharmacol.256, 201.
Cody, W.L., et al. 1993. Med. Chem. Res. 3, 154.
Doherty, A.M., et al. 1993. J. Cardiovasc.Pharmacol. 22, S98 (Suppl.).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service