Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09705

Sigma-Aldrich

L-Serine-O-phosphate

Synonym(s):

L-Serine-O-phosphate, L-2-Amino-3-hydroxypropanoic acid 3-phosphate, L-SOP, L-Serine O-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8NO6P
CAS Number:
Molecular Weight:
185.07
MDL number:
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 50 mg/mL

storage temp.

10-30°C

InChI

1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1

InChI key

BZQFBWGGLXLEPQ-REOHCLBHSA-N

General description

A potent agonist for group III mGlu receptor subtype 4, 6, and 8 (EC50 = 1-4, 3, and 2 µM for mGlu4, mGlu6, mGlu8, respectively); but not for subtype 7 (EC50 = 160-1200 µM). Inhibits proliferation and enhances neuronal differentiation in progenitor cells. Widely used to study presynaptic inhibitory modulation of the excitatory glutamatergic transmission at retina and central nervous systems.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
mGlu₄, mGlu₆, mGlu₈

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Eriksen, L., et al. 1995. Br. J. Pharmacol.116, 3279.

Thoreson, W., et al. 1995. Brain Res.676, 93.

Bruno, V., et al. 1996. Eur. J. Pharmacol.310, 61.
Saxe, J., et al. 2007. Chem. Biol.14, 1019.
Nakajima, Y., 1993. J. Biol. Chem.268, 11868.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service