Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09667

Sigma-Aldrich

VprBP Inhibitor, B32B3

Synonym(s):

VprBP Inhibitor, B32B3, Vpr Binding protein Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N5S
CAS Number:
Molecular Weight:
347.44
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

potency

500 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable, potent, selective, and ATP-competitive inhibitor of Vpr (HIV-1) binding protein (VprBP) that preferentially targets cancer cells expressing VprBP and blocks phosphorylation of H2A at Thr120 (IC50 = 500 nM in DU-145 human prostate cancer cells) and reduces cell proliferation. However, it has minimal effect on normal human prostate MLC cells lacking VprBP. Exhibits over 100-fold greater selectivity for VprBP over a panel of 33 other protein kinases. Shown to reduce xenograft tumor progression in nude mice injected with DU-145 cells (~5 mg/kg, i.p. twice a week for 3 weeks). Displays moderately desirable pharmacokinetic properties with t1/2= 7 h and Cmax of 1 µM.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
VprBP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, K., et al. 2013. Mol. Cell.52, 459.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service