Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.09266

Sigma-Aldrich

Mcl-1 Inhibitor III, UMI-77

Synonym(s):

Mcl-1 Inhibitor III, UMI-77, Myeloid Cell Leukemia-1 Inhibitor III, UMI77, 2-(4-(4-Bromophenylsulfonamido)-1-hydroxynaphthalen-2-ylthio)acetic acid, ((4-(((4-Bromophenyl)sulfonyl)amino)-1-hydroxy-2-naphthyl)sulfanyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14BrNO5S2
CAS Number:
Molecular Weight:
468.34
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable hydroxynaphthalenylthioacetate that exhibits Mcl-1-selective affinity (Ki against 2 nM FITC-BID79-99 binding = 490 nM/90 nM MCl-1 & 23.83 µM/60 nM Bcl-2; Ki against 1 nM FAM-BID80-99 binding = 5.33 µM/4 nM A1/Bfl-1; Ki against 2 nM FAM-BID80-99 binding = 8.19 µM/40 nM Bcl-w, 23.83 µM/60 nM Bcl-2, and 32.99 µM/50 nM Bcl-xL), effectively inhibiting Mcl-1 BH3-binding groove-mediated interactions with various binding partners, including Noxa, Bax, and Bak, by simultaneously targeting the groove′s h2 & h3 hydrophobic pockets. Shown to inhibit the proliferation of BxPC-3, Panc-1, and Capan-2 prostate cancer cultures (IC50 = 3.4, 4.4, and 5.5 µM, respectively) via apoptosis induction, while being less potent toward MiaPaca and AsPC-1 lines (IC50 = 12.5 and 16.1 µM, respectively) with high Bcl-xL and low Mcl-1/Bak expression and. Despite its moderate in vitro murine liver microsome stability (t1/2 = 45 mins), daily i.v. injection (60 mg/kg; 5 d/wk) is reported to effectively suppress BxPC-3 tumor expansion in mice in vivo without apparent animal toxicity, although severe weight loss is observed among mice received higher daily MUI-77 i.v. dosage of 80 mg/kg.
A cell-permeable, bioavailable hydroxynaphthalenylthioacetate compound that selectively and reversibly binds to the BH3 binding groove of Mcl-1 (Ki = 490 nM) and antagonizes its function by disrupting the heterodimerization of Mcl-1/Bax (IC50 = 1.43 µM) and Mcl-1/Bak in cells. Exhibits high selectivity over other members of the Bcl-2 family (Ki = 5.33, 8.19, 23.83, and 32.99 µM, for A1/Bfl-1, Bcl-w, Bcl-2, and Bcl-xL, respectively). Inhibits the growth of BxPC-3, Panc-1 cells, and Capan-2 cells (IC50 = 3.4, 4.4, and 5.5 µM, respectively) by inducing intrinsic apoptosis as evidenced by cytochrome c release and caspase-3 activation. However, it does not induce apoptosis in normal cells. Blocks the growth of BxPC-3 xenograft in SCID mice (60 mg/kg i.v. for 5 days for 2 weeks) without any cytotoxic effects on surrounding normal tissue. Exhibits desirable microsomal stability (t1/2 = 45 min. in murine models).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
binds to the BH3 binding groove of Mcl-1

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Abulwerdi, F., et al. 2014. Mol. Cancer Ther.13, 565.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service